1-(4-Chlorophenyl)cyclopropanamine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AM-1632-5G AM-1632-1G
COBBAM-1632-5GEA 0 GBP
COBBAM-1632-5G COBBAM-1632-1G
1-(4-Chlorophenyl)cyclopropanamine hydrochloride
1-(4-Chlorophenyl)cyclopropanamine hydrochloride
Formula: C₉H₁₁Cl₂N
MW: 204.0963 g/mol
MDL Number: MFCD07995727
CAS Number: 1009102-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR