1-(4-Chlorophenyl)cyclopropanamine hydrochloride
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22839G1
AS22839G5
ACTIAS22839G1EA
99
GBP
ACTIAS22839G1
ACTIAS22839G5
1-(4-Chlorophenyl)cyclopropanamine hydrochloride
1-(4-Chlorophenyl)cyclopropanamine hydrochloride
Formula:
C₉H₁₁Cl₂N MW: 204.0963 g/mol |
MDL Number:
MFCD07995727 CAS Number: 1009102-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...