2-Methyl-3-phenylbenzoic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methyl-[1,1'-biphenyl]-3-carboxylic acid

COBBYA-8643-1GEA 0 GBP
COBBYA-8643-1G
2-Methyl-3-phenylbenzoic acid
2-Methyl-3-phenylbenzoic acid
Formula: C₁₄H₁₂O₂
MDL Number: MFCD11554177
CAS Number: 115363-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR