5-Amino-4-cyano-3-methyl-1-phenyl-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1327-5G HI-1327-1G
COBBHI-1327-5GEA 0 GBP
COBBHI-1327-5G COBBHI-1327-1G
5-Amino-4-cyano-3-methyl-1-phenyl-1H-pyrazole
5-Amino-4-cyano-3-methyl-1-phenyl-1H-pyrazole
Formula: C₁₁H₁₀N₄
MDL Number: MFCD00128285
CAS Number: 5346-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR