3-(4-Chlorophenyl)-1H-pyrazol-5-amine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Amino-3-(4-chlorophenyl)-1H-pyrazole, 5-(4-Chlorophenyl)-1H-pyrazol-3-amine

M-Clarity™ quality level = MQ100

646695-1G 646695-5G
SIAL646695-1GEA 59.3 GBP
SIAL646695-1G SIAL646695-5G
3-(4-Chlorophenyl)-1H-pyrazol-5-amine, Sigma-Aldrich®
3-(4-Chlorophenyl)-1H-pyrazol-5-amine
Formula: C₉H₈ClN₃
MW: 193.64 g/mol
MDL Number: MFCD00053046
CAS Number: 78583-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR