1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: O-(4-Fluorobenzyl)hydroxylamine hydrochloride

010009-1G 010009-5G
FLUO010009-1GEA 0 GBP
FLUO010009-1G FLUO010009-5G
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride
Formula: C₇H₉ClFNO
MW: 177.6039 g/mol
MDL Number: MFCD01114581
CAS Number: 51572-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR