1-(4-Bromophenylsulphonyl)-4-propylpiperazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0944-1G OT-0944-5G
COBBOT-0944-1GEA 0 GBP
COBBOT-0944-1G COBBOT-0944-5G
1-(4-Bromophenylsulphonyl)-4-propylpiperazine
1-(4-Bromophenylsulphonyl)-4-propylpiperazine
Formula: C₁₃H₁₉BrN₂O₂S
MDL Number: MFCD06029950
CAS Number: 888181-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR