Brefeldin A, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-oneBrefeldin A, natural

203729-1EA 172 GBP
203729-1
Brefeldin A, Millipore®
Brefeldin A
Formula: C₁₆H₂₄O₄
MW: 280.4 g/mol
MDL Number: MFCD00083258
CAS Number: 20350-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR