Estradiol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: beta-Estradiol17β-Estradiol

M-Clarity™ quality level = MQ200

SIALE2257-1MGEA 113 GBP
SIALE2257-1MG
Estradiol, Sigma-Aldrich®
Estradiol
Formula: C₁₈H₂₄O₂
MW: 272.39 g/mol
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR