(S)-(+)-1-Boc-3-hydroxypyrrolidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

AS20192G100 AS20192G25
ACTIAS20192G100EA 0 GBP
ACTIAS20192G100 ACTIAS20192G25
(S)-(+)-1-Boc-3-hydroxypyrrolidine
(S)-(+)-1-Boc-3-hydroxypyrrolidine
Formula: C₉H₁₇NO₃
MW: 187.2362 g/mol
MDL Number: MFCD01317839
CAS Number: 101469-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR