(±)-1-Boc-3-aminopyrrolidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Aminopyrrolidine, N1-BOC protected, 1-Boc-3-Aminopyrrolidine

AS22359G1 AS22359G25 AS22359G5
ACTIAS22359G1EA 0 GBP
ACTIAS22359G1 ACTIAS22359G25 ACTIAS22359G5
(±)-1-Boc-3-aminopyrrolidine
(±)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD01861220
CAS Number: 186550-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR