(R)-(+)-1-Boc-3-aminopyrrolidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-1-Boc-3-aminopyrrolidine, (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine, (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine

M-Clarity™ quality level = MQ100

644064-1G 644064-5G
SIAL644064-1GEA 53.9 GBP
SIAL644064-1G SIAL644064-5G
(R)-(+)-1-Boc-3-aminopyrrolidine, Sigma-Aldrich®
(R)-(+)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD03419272
CAS Number: 147081-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR