2-Amino-5-cyanopyrazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Aminopyrazine-2-carbonitrile

AS1215G1 AS1215G5
ACTIAS1215G1EA 44 GBP
ACTIAS1215G1 ACTIAS1215G5
2-Amino-5-cyanopyrazine
2-Amino-5-cyanopyrazine
Formula: C₅H₄N₄
MW: 120.1100 g/mol
MDL Number: MFCD08234954
CAS Number: 113305-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR