3-Amino-4-bromo-1H-pyrazole

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

CFF538.2.5 CFF538.10
CHMPCFF538.2.5EA 0 GBP
CHMPCFF538.2.5 CHMPCFF538.10
3-Amino-4-bromo-1H-pyrazole
3-Amino-4-bromo-1H-pyrazole
Formula: C₃H₄BrN₃
MW: 161.99 g/mol
MDL Number: MFCD00082728
CAS Number: 16461-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR