2-Quinolinol
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Quinolone
, Quinolin-2(1H)-one
, 2-Hydroxyquinoline
, Carbostyril
, Quinolin-2-ol
COBBPY-7946-5GEA
0
GBP
COBBPY-7946-5G
COBBPY-7946-1G
COBBPY-7946-25G
2-Quinolinol
2-Quinolinol
Formula:
C₉H₇NO MW: 145.16 g/mol Boiling Pt: 347 °C (1013 hPa) Melting Pt: 199…200 °C Density: 1.37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006743 CAS Number: 59-31-4 EINECS: 200-420-6 Merck Index: 13,01835 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...