Methyl indole-4-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS50094G1 AS50094G5
ACTIAS50094G1EA 0 GBP
ACTIAS50094G1 ACTIAS50094G5
Methyl indole-4-carboxylate
Methyl indole-4-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00191222
CAS Number: 39830-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR