4'-Butylacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4'-n-Butylacetophenone, 4-Butylacetophenone, p-Butylacetophenone

AS24919G100 AS24919G25
ACTIAS24919G100EA 106 GBP
ACTIAS24919G100 ACTIAS24919G25
4'-Butylacetophenone
4'-Butylacetophenone
Formula: C₁₂H₁₆O
MW: 176.26 g/mol
MDL Number: MFCD00017500
CAS Number: 37920-25-5
EINECS: 253-715-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR