4'-Bromovalerophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24921G100 AS24921G25
ACTIAS24921G100EA 0 GBP
ACTIAS24921G100 ACTIAS24921G25
4'-Bromovalerophenone
4'-Bromovalerophenone
Formula: C₁₁H₁₃BrO
MW: 241.13 g/mol
MDL Number: MFCD00000107
CAS Number: 7295-44-5
EINECS: 230-729-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR