5'-Fluoro-2'-hydroxyacetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(5-Fluoro-2-hydroxyphenyl)ethanone
AS20532G100
AS20532G25
ACTIAS20532G100EA
105
GBP
ACTIAS20532G100
ACTIAS20532G25
5'-Fluoro-2'-hydroxyacetophenone
5'-Fluoro-2'-hydroxyacetophenone
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Boiling Pt: 182 °C (1013 hPa) Melting Pt: 56 °C |
MDL Number:
MFCD00011668 CAS Number: 394-32-1 EINECS: 206-893-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...