(S)-(-)-Nicotine USP
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
(-)-Nicotine
, L-Nicotine
, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine
, L(-)-Nicotine
, (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine
, Nicotine
S12398.PGEA
0
GBP
S12398.PG
(S)-(-)-Nicotine USP
(S)-(-)-Nicotine
Formula:
C₁₀H₁₄N₂ MW: 162.23 g/mol Boiling Pt: 247 °C (1013 hPa) Melting Pt: –79 °C Density: 1.01 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006369 CAS Number: 54-11-5 EINECS: 200-193-3 UN: 1654 ADR: 6.1,II Merck Index: 12,06611 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...