4-Propoxy-3-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Methyl-4-propoxybenzeneboronic acid, (3-Methyl-4-propoxyphenyl)boronic acid

BB-2742-1G BB-2742-5G
COBBBB-2742-1GEA 0 GBP
COBBBB-2742-1G COBBBB-2742-5G
4-Propoxy-3-methylphenylboronic acid
4-Propoxy-3-methylphenylboronic acid
Formula: C₁₀H₁₅BO₃
MW: 194.0400 g/mol
MDL Number: MFCD06659872
CAS Number: 279262-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR