Bis(pentafluorophenyl)dimethylsilane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010237-1G
010237-5G
FLUO010237-1GEA
0
GBP
FLUO010237-1G
FLUO010237-5G
Bis(pentafluorophenyl)dimethylsilane
Bis(pentafluorophenyl)dimethylsilane
Formula:
C₁₄H₆F₁₀Si |
MDL Number:
MFCD00042066 CAS Number: 10536-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...