8-Methyl-2-quinolinol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
8-Methylquinolin-2(1H)-one, 8-Methyl-2(1H)-quinolinone
PY-1271-1G
PY-1271-5G
COBBPY-1271-1GEA
0
GBP
COBBPY-1271-1G
COBBPY-1271-5G
8-Methyl-2-quinolinol
8-Methyl-2-quinolinol
Formula:
C₁₀H₉NO MW: 159.1846 g/mol |
MDL Number:
MFCD13195790 CAS Number: 4053-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...