6-Methyl-2-quinolinol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1185-1G PY-1185-5G
COBBPY-1185-1GEA 0 GBP
COBBPY-1185-1G COBBPY-1185-5G
6-Methyl-2-quinolinol
6-Methyl-2-quinolinol
Formula: C₁₀H₉NO
MW: 159.1846 g/mol
MDL Number: MFCD08236767
CAS Number: 4053-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR