2-Bromo-4,5,6-trichloroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-2,3,4-trifluoroaniline, 2-Bromo-4,5,6-trifluoroaniline

005376-1G 005376-5G
FLUO005376-1GEA 0 GBP
FLUO005376-1G FLUO005376-5G
2-Bromo-4,5,6-trichloroaniline
2-Bromo-4,5,6-trichloroaniline
Formula: C₆H₃BrF₃N
MW: 225.9900 g/mol
MDL Number: MFCD00013388
CAS Number: 122375-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR