Kaempferol

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

ROTH5859.1EA 236.97 GBP
ROTH5859.1
Kaempferol
Kaempferol
Formula: C₁₅H₁₀O₆
MW: 286.24 g/mol
MDL Number: MFCD00016938
CAS Number: 520-18-3
EINECS: 208-287-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR