Kaempferol, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

420345-25EA 95 GBP
420345-25
Kaempferol, Millipore®
Kaempferol
Formula: C₁₅H₁₀O₆
MW: 286.24 g/mol
MDL Number: MFCD00016938
CAS Number: 520-18-3
EINECS: 208-287-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR