Kaempferol ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
J60373.MFEA
100
GBP
J60373.MF
Kaempferol ≥98%
Kaempferol
Inhibits topoisomerase I catalyzed DNA religation. Induces apoptosis and is cytotoxic to human leukemic cell lines
Formula:
C₁₅H₁₀O₆ MW: 286.24 g/mol Melting Pt: 276…278 °C |
MDL Number:
MFCD00016938 CAS Number: 520-18-3 EINECS: 208-287-6 Merck Index: 14,5274 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...