4-Fluoro-3-(trifluoromethyl)benzophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006401-1G
006401-5G
006401-10G
FLUO006401-1GEA
0
GBP
FLUO006401-1G
FLUO006401-5G
FLUO006401-10G
4-Fluoro-3-(trifluoromethyl)benzophenone
4-Fluoro-3-(trifluoromethyl)benzophenone
Formula:
C₁₄H₈F₄O MW: 268.21 g/mol |
MDL Number:
MFCD00061258 CAS Number: 239087-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...