2-Fluoro-3-(trifluoromethyl)benzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006395-1G 006395-5G 006395-10G
FLUO006395-1GEA 0 GBP
FLUO006395-1G FLUO006395-5G FLUO006395-10G
2-Fluoro-3-(trifluoromethyl)benzophenone
2-Fluoro-3-(trifluoromethyl)benzophenone
Formula: C₁₄H₈F₄O
MW: 268.21 g/mol
MDL Number: MFCD00061151
CAS Number: 207853-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR