Vincristine sulphate 95%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
ACRO203440050EA
390
GBP
ACRO203440050
Vincristine sulphate 95%
Vincristine sulphate
Formula:
C₄₆H₅₈N₄O₁₄S MW: 923.04 g/mol Melting Pt: >300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00084729 CAS Number: 2068-78-2 EINECS: 218-190-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤12 % |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...