2-Bromo-4-chloro-5-fluoroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-Chloro-5-fluoroaniline

AS1608G1 AS1608G5
ACTIAS1608G1EA 0 GBP
ACTIAS1608G1 ACTIAS1608G5
2-Bromo-4-chloro-5-fluoroaniline
2-Bromo-4-chloro-5-fluoroaniline
Formula: C₆H₄BrClFN
MW: 224.458 g/mol
MDL Number: MFCD12026231
CAS Number: 120694-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR