3-Hydroxyisonicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Hydroxypyridine-4-carboxylic acid

AS30509G1 AS30509G5
ACTIAS30509G1EA 18 GBP
ACTIAS30509G1 ACTIAS30509G5
3-Hydroxyisonicotinic acid
3-Hydroxyisonicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00234165
CAS Number: 10128-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR