D-(+)-Cellobiose
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Cellose
, β-D-Glc-(1→4)-D-Glc
, 4-β-D-glucopyranosyl-D-glueopyranose
, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal
, β-Cellobiose
, 4-O-β-D-Glucopyranosyl-D-glucopyranose
EHERC11067000EA
107
GBP
EHERC11067000
D-(+)-Cellobiose
D(+)-Cellobiose
Formula:
C₁₂H₂₂O₁₁ MW: 342.30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00136034 CAS Number: 528-50-7 EINECS: 208-436-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...