5-Fluoro-2-methyl-4-nitroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21410G1 AS21410G5
ACTIAS21410G1EA 20 GBP
ACTIAS21410G1 ACTIAS21410G5
5-Fluoro-2-methyl-4-nitroaniline
5-Fluoro-2-methyl-4-nitroaniline
Formula: C₇H₇FN₂O₂
MW: 170.14 g/mol
MDL Number: MFCD07368827
CAS Number: 633327-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR