Erucic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(Z)-Docos-13-enoic acid
SIAL45630-1KG-FEA
179
GBP
SIAL45630-1KG-F
Erucic acid, Sigma-Aldrich®
Erucic acid
Formula:
C₂₂H₄₂O₂ MW: 338.58 g/mol |
MDL Number:
MFCD00063188 CAS Number: 112-86-7 EINECS: 204-011-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...