2-Bromo-p-anisidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-methoxyaniline

AS33773G1 AS33773G5
ACTIAS33773G1EA 17 GBP
ACTIAS33773G1 ACTIAS33773G5
2-Bromo-p-anisidine
2-Bromo-p-anisidine
Formula: C₇H₈BrNO
MW: 202.0485 g/mol
MDL Number: MFCD09029057
CAS Number: 32338-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR