4-Propoxyphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-n-Propoxybenzeneboronic acid

AS25098G25 AS25098G5
ACTIAS25098G25EA 179 GBP
ACTIAS25098G25 ACTIAS25098G5
4-Propoxyphenylboronic acid
4-Propoxyphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
MDL Number: MFCD03427189
CAS Number: 186497-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR