2,4-Dichlorobenzophenone ≥97.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D0342-5G
D0342-25G
TCIAD0342-5GEA
0
GBP
TCIAD0342-5G
TCIAD0342-25G
2,4-Dichlorobenzophenone ≥97.0% (by GC)
2,4-Dichlorobenzophenone
Formula:
C₁₃H₈Cl₂O |
MDL Number:
MFCD00018374 CAS Number: 19811-05-3 EINECS: 243-338-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 45.0 to 49.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...