6-Bromoquinolin-2(1H)-one

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7954-1G PY-7954-5G
COBBPY-7954-1GEA 0 GBP
COBBPY-7954-1G COBBPY-7954-5G
6-Bromoquinolin-2(1H)-one
6-Bromoquinolin-2(1H)-one
Formula: C₉H₆BrNO
MW: 224.05 g/mol
MDL Number: MFCD08703169
CAS Number: 1810-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR