6-Bromoquinolin-2(1H)-one
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS2240G1
AS2240G5
ACTIAS2240G1EA
0
GBP
ACTIAS2240G1
ACTIAS2240G5
6-Bromoquinolin-2(1H)-one
6-Bromoquinolin-2(1H)-one
Formula:
C₉H₆BrNO MW: 224.05 g/mol |
MDL Number:
MFCD08703169 CAS Number: 1810-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...