6-(2,4-Difluorophenyl)-3-pyridinecarbaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24611G1 AS24611G5
ACTIAS24611G1EA 274 GBP
ACTIAS24611G1 ACTIAS24611G5
6-(2,4-Difluorophenyl)-3-pyridinecarbaldehyde
6-(2,4-Difluorophenyl)-3-pyridinecarbaldehyde
Formula: C₁₂H₇F₂NO
MDL Number: MFCD04115483
CAS Number: 898796-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR