6-Chloronicotinoyl chloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20370G100 AS20370G25
ACTIAS20370G100EA 0 GBP
ACTIAS20370G100 ACTIAS20370G25
6-Chloronicotinoyl chloride
6-Chloronicotinoyl chloride
Formula: C₆H₃Cl₂NO
MW: 176.00 g/mol
MDL Number: MFCD00051775
CAS Number: 58757-38-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR