5-Bromo-2-chloronicotinamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1238-5G PY-1238-1G
COBBPY-1238-5GEA 0 GBP
COBBPY-1238-5G COBBPY-1238-1G
5-Bromo-2-chloronicotinamide
5-Bromo-2-chloronicotinamide
Formula: C₆H₄BrClN₂O
MDL Number: MFCD13176452
CAS Number: 75291-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR