6,8-Dimethylquinolin-2(1H)-one

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0213-5G IN-0213-1G
COBBIN-0213-5GEA 0 GBP
COBBIN-0213-5G COBBIN-0213-1G
6,8-Dimethylquinolin-2(1H)-one
6,8-Dimethylquinolin-2(1H)-one
Formula: C₁₁H₁₁NO
MDL Number: MFCD15143578
CAS Number: 54904-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR