4-Morpholino-2-pyridinamine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-4-(morpholin-4-yl)pyridine, 4-Morpholinopyridin-2-amine

AS22981G5 AS22981G1
ACTIAS22981G5EA 617 GBP
ACTIAS22981G5 ACTIAS22981G1
4-Morpholino-2-pyridinamine
4-Morpholino-2-pyridinamine
Formula: C₉H₁₃N₃O
MW: 179.2200 g/mol
MDL Number: MFCD09991750
CAS Number: 722549-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR