2-Ethoxy-3-formyl-5-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Ethoxy-3-formyl-5-methylphenyl)boronic acid

BB-8123-1G BB-8123-250M
COBBBB-8123-1GEA 0 GBP
COBBBB-8123-1G COBBBB-8123-250M
2-Ethoxy-3-formyl-5-methylphenylboronic acid
2-Ethoxy-3-formyl-5-methylphenylboronic acid
Formula: C₁₀H₁₃BO₄
MW: 208.0200 g/mol
MDL Number: MFCD08276749
CAS Number: 480424-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR