2'-(4-Fluorobenzyloxy)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010106-25G 010106-5G
FLUO010106-25GEA 0 GBP
FLUO010106-25G FLUO010106-5G
2'-(4-Fluorobenzyloxy)acetophenone
2'-(4-Fluorobenzyloxy)acetophenone
Formula: C₁₅H₁₃FO₂
MDL Number: MFCD03001269
CAS Number: 400878-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR