4-Fluoro-4'-nitrobenzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Fluorophenyl)(4-nitrophenyl)methanone

006298-5G 006298-10G 006298-1G
FLUO006298-5GEA 0 GBP
FLUO006298-5G FLUO006298-10G FLUO006298-1G
4-Fluoro-4'-nitrobenzophenone
4-Fluoro-4'-nitrobenzophenone
Formula: C₁₃H₈FNO₃
MW: 245.2059 g/mol
MDL Number: MFCD00053428
CAS Number: 2195-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR