4,5-Dimethoxy-2-nitrotoluene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,2-dimethoxy-4-methyl-5-nitrobenzene

D4641-F.5 D4641-F.25
XXXXD4641-F.5EA 0 GBP
XXXXD4641-F.5 XXXXD4641-F.25
4,5-Dimethoxy-2-nitrotoluene
4,5-Dimethoxy-2-nitrotoluene
Formula: C₉H₁₁NO₄
MW: 197.187 g/mol
MDL Number: MFCD00060708
CAS Number: 7509-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR